siRNA Details
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301140.1 | NP_001288069.1 | 6 | 1890 | ||
NM_001301141.1 | NP_001288070.1 | 6 | 1901 | |||
NM_001320042.1 | NP_001306971.1 | 5 | 2977 | |||
NM_014301.4 | NP_055116.1 | 6 | 1900 | |||
NM_213595.3 | NP_998760.1 | 5 | 1804 | |||
NR_135127.1 | 7 | 2000 | ||||
XM_011538100.2 | XP_011536402.1 | 6 | 3073 | |||
XM_011538101.2 | XP_011536403.1 | 4 | 1725 | |||
GenBank mRNA | AK000574.1 | - | 5 | 647 | ||
AK022758.1 | - | 4 | 3401 | |||
AK057251.1 | - | 6 | 1901 | |||
AY009127.1 | - | 6 | 755 | |||
AY009128.1 | - | 5 | 659 | |||
BC011906.2 | - | 5 | 660 | 4136262 | ||
BC061903.1 | - | 5 | 653 | 4183674 | ||
BG621612.1 | - | 271 | ||||
BG702203.1 | - | 6 | 741 | |||
BM423850.1 | - | 5 | 650 | |||
BM921073.1 | - | 5 | 745 | |||
EU329002.1 | - | 7 | 857 | |||
U47101.1 | - | 4 | 507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
