siRNA Details
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301140.1 | NP_001288069.1 | 3 | 1442 | ||
NM_001301141.1 | NP_001288070.1 | 3 | 1442 | |||
NM_001320042.1 | NP_001306971.1 | 3 | 1442 | |||
NM_014301.4 | NP_055116.1 | 4 | 1538 | |||
NM_213595.3 | NP_998760.1 | 3 | 1442 | |||
NR_135127.1 | 4 | 1538 | ||||
XM_011538100.2 | XP_011536402.1 | 4 | 1538 | |||
XM_011538101.2 | XP_011536403.1 | 3 | 1442 | |||
GenBank mRNA | AK000574.1 | - | 3 | 285 | ||
AK022758.1 | - | 2 | 1866 | |||
AK057251.1 | - | 3 | 1442 | |||
AK293202.1 | - | 3 | 295 | |||
AK297862.1 | - | 3 | 314 | |||
AY009127.1 | - | 4 | 395 | |||
AY009128.1 | - | 3 | 299 | |||
BC011906.2 | - | 3 | 298 | 4136262 | ||
BC061903.1 | - | 3 | 291 | 4183674 | ||
BG702203.1 | - | 3 | 293 | |||
BI597126.1 | - | 3 | 393 | |||
BM423850.1 | - | 3 | 288 | |||
BM921073.1 | - | 3 | 383 | |||
BP378051.1 | - | 3 | 314 | |||
BU602914.1 | - | 3 | 303 | |||
DB095495.1 | - | 3 | 309 | |||
DB493673.1 | - | 3 | 372 | |||
EU329002.1 | - | 4 | 395 | |||
U47101.1 | - | 2 | 145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
