siRNA Details
Gene Symbol: ACAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135.3 | NP_001126.3 | 2, 3 | 443 | ||
NM_013227.3 | NP_037359.3 | 2, 3 | 443 | |||
XM_006720419.1 | XP_006720482.1 | 2, 3 | 444 | |||
XM_011521313.1 | XP_011519615.1 | 2, 3 | 444 | |||
XM_011521314.1 | XP_011519616.1 | 2, 3 | 444 | |||
XM_017021985.1 | XP_016877474.1 | 2, 3 | 444 | |||
XM_017021986.1 | XP_016877475.1 | 2, 3 | 444 | |||
XM_017021987.1 | XP_016877476.1 | 2, 3 | 444 | |||
GenBank mRNA | AK290723.1 | - | 2, 3 | 443 | ||
BC036445.1 | - | 2, 3 | 414 | |||
BC150624.1 | - | 127 | 9056994 | |||
DA596050.1 | - | 2, 3 | 443 | |||
M55172.1 | - | 2, 3 | 129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
