siRNA Details
Gene Symbol: TMEM26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178505.7 | NP_848600.2 | 6 | 1198 | ||
NR_134507.1 | 6 | 1198 | ||||
GenBank mRNA | AK124367.1 | - | 2 | 1189 | ||
AK125611.1 | - | 6 | 986 | |||
CR749606.1 | - | 6 | 1198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
