siRNA Details
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164281.2 | NP_001157753.1 | 7, 8 | 1982 | ||
NM_001164283.2 | NP_001157755.1 | 2, 3 | 264 | |||
NM_147200.2 | NP_671733.2 | 8, 9 | 1773 | |||
NM_147686.3 | NP_679211.2 | 7, 8 | 1985 | |||
XM_006715319.3 | XP_006715382.1 | 2, 3 | 226 | |||
XM_011535386.1 | XP_011533688.1 | 8, 9 | 2708 | |||
GenBank mRNA | AF136405.1 | - | 8, 9 | 1681 | ||
AF136406.1 | - | 7, 8 | 1772 | |||
AF136407.1 | - | 7, 8 | 1772 | |||
AF272151.1 | - | 6, 7 | 1505 | |||
AF274303.1 | - | 7, 8 | 1505 | |||
AK025351.1 | - | 7, 8 | 1579 | |||
AK026602.1 | - | 6, 7 | 979 | |||
AK314415.1 | - | 8, 9 | 1696 | |||
AL050289.1 | - | 7, 8 | 1657 | |||
BC002823.2 | - | 7, 8 | 1568 | 3634505 | ||
BC026723.1 | - | 2, 3 | 183 | |||
BI856094.1 | - | 2, 3 | 262 | |||
BM449775.1 | - | 4, 5 | 447 | |||
CB049654.1 | - | 2, 3 | 231 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
