siRNA Details
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164281.2 | NP_001157753.1 | 7 | 1908 | ||
NM_001164283.2 | NP_001157755.1 | 2 | 190 | |||
NM_147200.2 | NP_671733.2 | 8 | 1699 | |||
NM_147686.3 | NP_679211.2 | 7 | 1911 | |||
XM_006715319.3 | XP_006715382.1 | 2 | 152 | |||
XM_011535386.1 | XP_011533688.1 | 8 | 2634 | |||
GenBank mRNA | AF136405.1 | - | 8 | 1607 | ||
AF136406.1 | - | 7 | 1698 | |||
AF136407.1 | - | 7 | 1698 | |||
AF272151.1 | - | 6 | 1431 | |||
AF274303.1 | - | 7 | 1431 | |||
AK025351.1 | - | 7 | 1505 | |||
AK026602.1 | - | 6 | 905 | |||
AK314415.1 | - | 8 | 1622 | |||
AL050289.1 | - | 7 | 1583 | |||
BC002823.2 | - | 7 | 1494 | 3634505 | ||
BC026723.1 | - | 2 | 109 | |||
BI856094.1 | - | 2 | 188 | |||
BM449775.1 | - | 4 | 373 | |||
CB049654.1 | - | 2 | 157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
