siRNA Details
Gene Symbol: PPP1R27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007533.3 | NP_001007534.1 | 3 | 763 | ||
GenBank mRNA | AF434846.1 | - | 3 | 741 | ||
AK096888.1 | - | 2 | 1413 | |||
BI826571.1 | - | 2 | 384 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
