siRNA Details
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007240.2 | NP_001007241.2 | 8 | 1275 | ||
NM_001007241.2 | NP_001007242.2 | 7 | 834 | |||
NM_001007242.2 | NP_001007243.2 | 6 | 825 | |||
NM_001502.3 | NP_001493.2 | 7 | 1266 | |||
XM_017023155.1 | XP_016878644.1 | 7 | 1019 | |||
GenBank mRNA | AB035541.1 | - | 6 | 1055 | ||
AB035542.1 | - | 5 | 614 | |||
AK293384.1 | - | 7 | 1022 | |||
AK312277.1 | - | 7 | 1153 | |||
BC032693.2 | - | 5 | 672 | 5229838 | ||
BX647831.1 | - | 8 | 1163 | |||
BX648023.1 | - | 7 | 1160 | |||
BX648201.1 | - | 6 | 712 | |||
BX649067.1 | - | 7 | 721 | |||
CR749600.1 | - | 7 | 1225 | |||
D38225.1 | - | 6 | 626 | |||
U36221.1 | - | 7 | 1135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
