siRNA Details
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007240.2 | NP_001007241.2 | 12 | 1937 | ||
NM_001007241.2 | NP_001007242.2 | 11 | 1496 | |||
NM_001007242.2 | NP_001007243.2 | 10 | 1487 | |||
NM_001502.3 | NP_001493.2 | 11 | 1928 | |||
XM_017023155.1 | XP_016878644.1 | 11 | 1681 | |||
GenBank mRNA | AK293384.1 | - | 11 | 1684 | ||
BC032693.2 | - | 9 | 1334 | 5229838 | ||
BX647831.1 | - | 12 | 1825 | |||
BX648023.1 | - | 11 | 1822 | |||
BX649067.1 | - | 11 | 1383 | |||
CR749600.1 | - | 11 | 1887 | |||
U36221.1 | - | 11 | 1797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
