siRNA Details
Gene Symbol: CCDC62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201435.4 | NP_958843.2 | 4 | 627 | ||
NR_027918.2 | 4 | 627 | ||||
XM_006719643.1 | XP_006719706.1 | 4 | 535 | |||
XM_006719644.1 | XP_006719707.1 | 4 | 535 | |||
XM_011538856.1 | XP_011537158.1 | 4 | 535 | |||
XM_011538857.1 | XP_011537159.1 | 4 | 535 | |||
GenBank mRNA | AK097663.1 | - | 4 | 551 | ||
AK292466.1 | - | 4 | 535 | |||
AY254201.1 | - | 4 | 608 | |||
BC060796.1 | - | 4 | 610 | 5284906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
