siRNA Details
Gene Symbol: CNOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265612.1 | NP_001252541.1 | 43, 44 | 6605 | ||
NM_016284.4 | NP_057368.3 | 43, 44 | 6620 | |||
NR_049763.1 | 43, 44 | 6605 | ||||
GenBank mRNA | AB023224.2 | - | 30, 31 | 4665 | ||
AF110778.1 | - | 3, 4 | 397 | |||
AK056927.1 | - | 20, 21 | 2950 | |||
AK075125.1 | - | 13, 14 | 1919 | |||
AK092885.1 | - | 3, 4 | 1164 | |||
AK301092.1 | - | 3, 4 | 384 | |||
AK302495.1 | - | 20, 21 | 2990 | |||
AL117492.1 | - | 15, 16 | 2459 | |||
AL832769.1 | - | 43, 44 | 6569 | |||
AL833549.1 | - | 43, 44 | 6579 | |||
BC000779.2 | - | 5, 6 | 778 | |||
BC024317.1 | - | 26, 27 | 4019 | |||
BC027969.2 | - | 2, 3 | 234 | |||
BX537840.1 | - | 43, 44 | 6551 | |||
CR749237.1 | - | 36, 37 | 5560 | |||
EF553522.1 | - | 43, 44 | 6554 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
