siRNA Details
Gene Symbol: HS6ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004807.2 | NP_004798.3 | 2 | 1659 | ||
GenBank mRNA | AB006179.1 | - | 2 | 1733 | ||
BC001196.1 | - | 1 | 908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
