siRNA Details
Gene Symbol: SEC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136026.2 | NP_001129498.1 | 7 | 818 | ||
NM_001136232.2 | NP_001129704.1 | 6 | 672 | |||
NM_030673.3 | NP_109598.2 | 7 | 674 | |||
NM_183352.2 | NP_899195.1 | 6 | 640 | |||
XM_005265379.2 | XP_005265436.1 | 7 | 691 | |||
XM_017007019.1 | XP_016862508.1 | 7 | 728 | |||
XM_017007020.1 | XP_016862509.1 | 6 | 535 | |||
XM_017007021.1 | XP_016862510.1 | 6 | 545 | |||
GenBank mRNA | AB209554.1 | - | 5 | 487 | ||
AF052155.1 | - | 6 | 672 | |||
AK095629.1 | - | 4 | 2499 | |||
AK223019.1 | - | 6 | 525 | |||
AK301999.1 | - | 7 | 742 | |||
BC002634.2 | - | 7 | 555 | 3605545 | ||
BC006167.1 | - | 3 | 332 | |||
BC091506.1 | - | 6 | 550 | 30408210 | ||
CN344033.1 | - | 6 | 652 | |||
EF036494.1 | - | 6 | 539 | |||
L09260.1 | - | 6 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
