siRNA Details
Gene Symbol: SLC25A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006641.3 | NP_001006642.1 | 10 | 3001 | ||
NM_001006642.3 | NP_001006643.1 | 10 | 3246 | |||
NM_001265614.2 | NP_001252543.1 | 11 | 3282 | |||
NM_052901.4 | NP_443133.2 | 10 | 2864 | |||
NR_049766.2 | 10 | 2907 | ||||
XM_005251688.1 | XP_005251745.1 | 11 | 3037 | |||
XM_005251689.4 | XP_005251746.1 | 10 | 3260 | |||
XM_006716948.2 | XP_006717011.1 | 11 | 3438 | |||
XM_006716949.3 | XP_006717012.1 | 11 | 3298 | |||
GenBank mRNA | AB067483.1 | - | 11 | 3281 | ||
AJ619963.1 | - | 10 | 3165 | |||
AJ619989.1 | - | 10 | 2789 | |||
AJ619990.1 | - | 10 | 2884 | |||
AJ619991.1 | - | 10 | 2846 | |||
AJ619992.1 | - | 10 | 2832 | |||
AY358515.1 | - | 10 | 2839 | |||
BC005163.2 | - | 6 | 2298 | |||
BC044631.1 | - | 1 | 1424 | |||
BC061925.1 | - | 1 | 421 | |||
BC089448.1 | - | 10 | 2855 | 30558551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
