siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 27 | 5015 | ||
XM_011544278.2 | XP_011542580.2 | 20 | 2626 | |||
XM_017002391.1 | XP_016857880.1 | 26 | 4579 | |||
XM_017002392.1 | XP_016857881.1 | 14 | 1865 | |||
GenBank mRNA | AB007895.1 | - | 4 | 1931 | ||
AK055374.1 | - | 8 | 1181 | |||
AK074078.1 | - | 22 | 4087 | |||
AK090784.1 | - | 3 | 1247 | |||
AK097229.1 | - | 13 | 1590 | |||
AK307812.1 | - | 14 | 1861 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
