siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 31 | 5666 | ||
XM_011544278.2 | XP_011542580.2 | 24 | 3277 | |||
XM_017002391.1 | XP_016857880.1 | 30 | 5230 | |||
XM_017002392.1 | XP_016857881.1 | 18 | 2516 | |||
GenBank mRNA | AB007895.1 | - | 8 | 2582 | ||
AK055374.1 | - | 12 | 1832 | |||
AK074078.1 | - | 26 | 4738 | |||
AK090784.1 | - | 7 | 1898 | |||
AK097229.1 | - | 17 | 2241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
