siRNA Details
Gene Symbol: PCNX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014801.3 | NP_055616.3 | 24 | 4351 | ||
XM_011544278.2 | XP_011542580.2 | 17 | 1962 | |||
XM_017002391.1 | XP_016857880.1 | 23 | 3915 | |||
XM_017002392.1 | XP_016857881.1 | 11 | 1201 | |||
GenBank mRNA | AB007895.1 | - | 1 | 1267 | ||
AK055374.1 | - | 5 | 517 | |||
AK074078.1 | - | 19 | 3423 | |||
AK097229.1 | - | 11 | 1180 | |||
AK307812.1 | - | 11 | 1197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
