siRNA Details
Gene Symbol: ISM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182509.3 | NP_872315.2 | 2 | 308 | ||
NM_199296.2 | NP_954993.1 | 2 | 308 | |||
XM_011536489.1 | XP_011534791.1 | 2 | 198 | |||
GenBank mRNA | AJ583024.1 | - | 2 | 269 | ||
AK056709.1 | - | 3 | 362 | |||
AK075445.1 | - | 2 | 291 | |||
AK291600.1 | - | 2 | 257 | |||
AK300214.1 | - | 2 | 257 | |||
BC033140.1 | - | 1 | 75 | |||
BC101019.1 | - | 2 | 257 | 40007143 | ||
BC101020.1 | - | 2 | 257 | 40007145 | ||
BC101021.2 | - | 2 | 257 | 40007146 | ||
BC101022.2 | - | 2 | 257 | 40007147 | ||
BX248277.1 | - | 2 | 241 | |||
BX248770.1 | - | 2 | 247 | |||
BX248771.1 | - | 2 | 628 | |||
CR749863.1 | - | 2 | 257 | |||
DB025298.1 | - | 2 | 309 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
