siRNA Details
Gene Symbol: ISM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182509.3 | NP_872315.2 | 5, 6 | 934 | ||
NM_199296.2 | NP_954993.1 | 6, 7 | 1280 | |||
XM_011536489.1 | XP_011534791.1 | 6, 7 | 1170 | |||
GenBank mRNA | AJ583024.1 | - | 6, 7 | 1241 | ||
AK056709.1 | - | 7, 8 | 1334 | |||
AK075445.1 | - | 6, 7 | 1263 | |||
AK291600.1 | - | 6, 7 | 1229 | |||
AK300214.1 | - | 5, 6 | 986 | |||
BC033140.1 | - | 5, 6 | 1047 | |||
BC101019.1 | - | 6, 7 | 1229 | 40007143 | ||
BC101020.1 | - | 6, 7 | 1229 | 40007145 | ||
BC101021.2 | - | 5, 6 | 883 | 40007146 | ||
BC101022.2 | - | 6, 7 | 1229 | 40007147 | ||
BX248277.1 | - | 5, 6 | 867 | |||
BX248280.1 | - | 4, 5 | 748 | |||
CR749863.1 | - | 6, 7 | 1229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
