siRNA Details
Gene Symbol: PAX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000280.4 | NP_000271.1 | 10 | 1415 | ||
NM_001127612.1 | NP_001121084.1 | 10 | 1329 | |||
NM_001258462.1 | NP_001245391.1 | 11 | 1371 | |||
NM_001258463.1 | NP_001245392.1 | 11 | 1309 | |||
NM_001258464.1 | NP_001245393.1 | 10 | 1317 | |||
NM_001258465.1 | NP_001245394.1 | 9 | 1303 | |||
NM_001310158.1 | NP_001297087.1 | 11 | 1412 | |||
NM_001310159.1 | NP_001297088.1 | 8 | 988 | |||
NM_001310160.1 | NP_001297089.1 | 7 | 2626 | |||
NM_001310161.1 | NP_001297090.1 | 8 | 1178 | |||
NM_001604.5 | NP_001595.2 | 11 | 1359 | |||
GenBank mRNA | AB209177.1 | - | 8 | 5767 | ||
AB593092.1 | - | 11 | 1310 | |||
AB593093.1 | - | 11 | 1309 | |||
AB593094.1 | - | 11 | 1412 | |||
AK074881.1 | - | 9 | 1750 | |||
AK094172.1 | - | 5 | 2053 | |||
AK094249.1 | - | 7 | 2436 | |||
AK314470.1 | - | 10 | 1307 | |||
AY047583.1 | - | 8 | 990 | |||
AY707088.1 | - | 6 | 860 | |||
BC011953.1 | - | 10 | 1201 | 3880468 | ||
BI816814.1 | - | 1 | 83 | |||
BX640762.1 | - | 11 | 1338 | |||
GQ141695.1 | - | 7 | 908 | |||
M77844.1 | - | 9 | 1222 | |||
M93650.1 | - | 10 | 1277 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
