siRNA Details
Gene Symbol: PAX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000280.4 | NP_000271.1 | 6 | 901 | ||
NM_001127612.1 | NP_001121084.1 | 6 | 815 | |||
NM_001258462.1 | NP_001245391.1 | 7 | 857 | |||
NM_001258463.1 | NP_001245392.1 | 7 | 795 | |||
NM_001258464.1 | NP_001245393.1 | 6 | 803 | |||
NM_001258465.1 | NP_001245394.1 | 5 | 789 | |||
NM_001310158.1 | NP_001297087.1 | 7 | 898 | |||
NM_001310159.1 | NP_001297088.1 | 4 | 474 | |||
NM_001310160.1 | NP_001297089.1 | 3 | 1781 | |||
NM_001310161.1 | NP_001297090.1 | 4 | 664 | |||
NM_001604.5 | NP_001595.2 | 7 | 845 | |||
GenBank mRNA | AB209177.1 | - | 4 | 5253 | ||
AB593092.1 | - | 7 | 796 | |||
AB593093.1 | - | 7 | 795 | |||
AB593094.1 | - | 7 | 898 | |||
AK074881.1 | - | 5 | 1236 | |||
AK094172.1 | - | 2 | 835 | |||
AK094249.1 | - | 3 | 1591 | |||
AK314470.1 | - | 6 | 793 | |||
AY047583.1 | - | 4 | 476 | |||
AY707088.1 | - | 2 | 346 | |||
BC011953.1 | - | 6 | 687 | 3880468 | ||
BX640762.1 | - | 7 | 824 | |||
DA571138.1 | - | 4 | 486 | |||
GQ141695.1 | - | 3 | 394 | |||
KP255960.1 | - | 3 | 388 | |||
M77844.1 | - | 5 | 708 | |||
M93650.1 | - | 6 | 763 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
