siRNA Details
Gene Symbol: SIGLEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527533.2 | XP_011525835.1 | 8 | 1997 | ||
XM_011527535.2 | XP_011525837.1 | 8 | 1911 | |||
XM_011527536.2 | XP_011525838.1 | 8 | 1836 | |||
XM_011527538.2 | XP_011525840.1 | 6 | 1823 | |||
GenBank mRNA | BC035359.1 | - | 7 | 1777 | 5176194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
