siRNA Details
Gene Symbol: SIGLEC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011527533.2 | XP_011525835.1 | 8 | 1524 | ||
XM_011527534.2 | XP_011525836.1 | 9 | 1633 | |||
XM_011527535.2 | XP_011525837.1 | 8 | 1438 | |||
XM_011527536.2 | XP_011525838.1 | 8 | 1363 | |||
XM_011527537.2 | XP_011525839.1 | 8 | 1330 | |||
XM_011527538.2 | XP_011525840.1 | 6 | 1350 | |||
XM_011527539.2 | XP_011525841.1 | 4 | 1010 | |||
GenBank mRNA | AK300170.1 | - | 7 | 1283 | ||
AK300182.1 | - | 7 | 1380 | |||
BC035359.1 | - | 7 | 1304 | 5176194 | ||
BC140798.1 | - | 6 | 1093 | 9021666 | ||
D86358.1 | - | 8 | 1486 | |||
D86359.1 | - | 6 | 1310 | |||
U71382.1 | - | 8 | 1280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
