siRNA Details
Gene Symbol: NCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000615.6 | NP_000606.3 | 8 | 1414 | ||
NM_001076682.3 | NP_001070150.1 | 8 | 1414 | |||
NM_001242607.1 | NP_001229536.1 | 8 | 1414 | |||
NM_001242608.1 | NP_001229537.1 | 8 | 1414 | |||
NM_181351.4 | NP_851996.2 | 8 | 1414 | |||
GenBank mRNA | AB209443.1 | - | 8 | 1293 | ||
AK057509.1 | - | 8 | 1517 | |||
AK292453.1 | - | 8 | 1049 | |||
AK314589.1 | - | 8 | 1386 | |||
BC014205.2 | - | 2 | 278 | |||
BC029119.1 | - | 8 | 1225 | |||
BC047244.1 | - | 8 | 1226 | 5590188 | ||
S71824.1 | - | 8 | 1249 | |||
U63041.1 | - | 8 | 1085 | |||
X16841.1 | - | 8 | 1193 | |||
X59400.1 | - | 1 | 28 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
