siRNA Details
Gene Symbol: GYPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308187.1 | NP_001295116.1 | 5, 6 | 543 | ||
NM_001308190.1 | NP_001295119.1 | 5, 6 | 483 | |||
NM_002099.7 | NP_002090.4 | 6, 7 | 582 | |||
XM_017008135.1 | XP_016863624.1 | 7, 8 | 649 | |||
GenBank mRNA | AK290561.1 | - | 6, 7 | 547 | ||
AY950614.1 | - | 6, 7 | 431 | |||
BC005319.1 | - | 6, 7 | 488 | 3930700 | ||
BC013328.1 | - | 6, 7 | 469 | 4134368 | ||
BG497403.1 | - | 1, 2 | 27 | |||
EU338231.1 | - | 7, 8 | 551 | |||
EU338232.1 | - | 6, 7 | 468 | |||
EU338233.1 | - | 7, 8 | 551 | |||
EU338234.1 | - | 5, 6 | 375 | |||
GU347002.1 | - | 7, 8 | 575 | |||
GU347003.1 | - | 7, 8 | 654 | |||
GU347004.1 | - | 7, 8 | 632 | |||
GU347005.1 | - | 2, 3 | 154 | |||
GU347007.1 | - | 3, 4 | 234 | |||
L07251.1 | - | 335 | ||||
L07253.1 | - | 296 | ||||
L31860.1 | - | 6, 7 | 486 | |||
M12857.1 | - | 6, 7 | 441 | |||
U00177.1 | - | 6, 7 | 431 | |||
U00178.1 | - | 5, 6 | 335 | |||
U00179.1 | - | 4, 5 | 236 | |||
X08054.1 | - | 6, 7 | 487 | |||
X51798.1 | - | 6, 7 | 455 | |||
X57023.1 | - | 5, 6 | 392 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
