siRNA Details
Gene Symbol: IP6K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005909.2 | NP_001005909.1 | 3 | 592 | ||
NM_016291.3 | NP_057375.2 | 3 | 595 | |||
XM_006713199.3 | XP_006713262.1 | 4 | 699 | |||
XM_006713200.1 | XP_006713263.1 | 4 | 669 | |||
XM_006713201.1 | XP_006713264.1 | 4 | 746 | |||
XM_006713202.1 | XP_006713265.1 | 4 | 748 | |||
XM_011533816.1 | XP_011532118.1 | 4 | 729 | |||
XM_011533817.1 | XP_011532119.1 | 5 | 1634 | |||
XM_011533818.1 | XP_011532120.1 | 4 | 1902 | |||
XM_017006583.1 | XP_016862072.1 | 3 | 591 | |||
XM_017006584.1 | XP_016862073.1 | 3 | 566 | |||
XM_017006585.1 | XP_016862074.1 | 4 | 681 | |||
XM_017006586.1 | XP_016862075.1 | 4 | 2542 | |||
XM_017006587.1 | XP_016862076.1 | 7 | 1891 | |||
GenBank mRNA | AF177145.1 | - | 2 | 489 | ||
AK315204.1 | - | 3 | 533 | |||
AL117458.1 | - | 1 | 119 | |||
AY007091.1 | - | 3 | 518 | |||
BC019694.1 | - | 3 | 504 | 4941646 | ||
BC065533.1 | - | 3 | 521 | 6162048 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
