siRNA Details
Gene Symbol: DHX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001357.4 | NP_001348.2 | 13 | 1546 | ||
NR_033302.1 | 14 | 1683 | ||||
GenBank mRNA | AK226102.1 | - | 14 | 1664 | ||
AK308165.1 | - | 13 | 1500 | |||
BC008773.1 | - | 13 | 1463 | |||
BC014246.2 | - | 13 | 1481 | |||
BC025245.1 | - | 13 | 1468 | |||
BC137136.1 | - | 13 | 1493 | 9021133 | ||
BC144494.1 | - | 13 | 1493 | |||
L13848.1 | - | 13 | 1451 | |||
Y10658.1 | - | 12 | 1371 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
