siRNA Details
Gene Symbol: DHX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001357.4 | NP_001348.2 | 24 | 2989 | ||
NR_033302.1 | 25 | 3126 | ||||
GenBank mRNA | AK096582.1 | - | 5 | 1219 | ||
AK226102.1 | - | 25 | 3107 | |||
BC058896.1 | - | 3 | 212 | |||
BC107881.1 | - | 4 | 333 | |||
BC137136.1 | - | 24 | 2936 | 9021133 | ||
BC144494.1 | - | 23 | 2862 | |||
L13848.1 | - | 24 | 2891 | |||
U03643.1 | - | 2 | 171 | |||
Y10658.1 | - | 23 | 2814 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
