siRNA Details
Gene Symbol: ABHD12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206673.1 | NP_001193602.1 | 7 | 571 | ||
NM_181533.3 | NP_853511.2 | 6 | 443 | |||
NM_181814.1 | NP_861535.1 | 5 | 340 | |||
XM_006720052.1 | XP_006720115.1 | 5 | 337 | |||
XM_011536472.1 | XP_011534774.1 | 7 | 641 | |||
XM_011536474.2 | XP_011534776.1 | 7 | 1092 | |||
XM_017021031.1 | XP_016876520.1 | 6 | 505 | |||
GenBank mRNA | AK128464.1 | - | 5 | 337 | ||
AY311396.1 | - | 7 | 571 | |||
AY311397.1 | - | 6 | 443 | |||
AY311398.1 | - | 5 | 340 | |||
BC034603.1 | - | 5 | 307 | 4778051 | ||
BC107141.1 | - | 5 | 340 | 40030548 | ||
BC107142.1 | - | 5 | 338 | 40030549 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
