siRNA Details
Gene Symbol: ABHD12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206673.1 | NP_001193602.1 | 13 | 1568 | ||
NM_181533.3 | NP_853511.2 | 12 | 1440 | |||
NM_181814.1 | NP_861535.1 | 11 | 1337 | |||
XM_011536474.2 | XP_011534776.1 | 13 | 2089 | |||
GenBank mRNA | AY311396.1 | - | 13 | 1568 | ||
AY311397.1 | - | 12 | 1440 | |||
AY311398.1 | - | 11 | 1337 | |||
AY311399.1 | - | 10 | 1264 | |||
BC034603.1 | - | 11 | 1304 | 4778051 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
