siRNA Details
Gene Symbol: CHIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040623.2 | NP_001035713.1 | 10 | 1004 | ||
NM_001258001.1 | NP_001244930.1 | 11 | 1170 | |||
NM_001258002.1 | NP_001244931.1 | 8 | 881 | |||
NM_001258003.1 | NP_001244932.1 | 10 | 1077 | |||
NM_001258004.1 | NP_001244933.1 | 12 | 1309 | |||
NM_001258005.1 | NP_001244934.1 | 11 | 1143 | |||
NM_021797.3 | NP_068569.2 | 9 | 1047 | |||
NM_201653.3 | NP_970615.2 | 12 | 1372 | |||
XM_006710577.3 | XP_006710640.1 | 9 | 1039 | |||
XM_017001047.1 | XP_016856536.1 | 7 | 892 | |||
XM_017001048.1 | XP_016856537.1 | 10 | 1080 | |||
GenBank mRNA | AB025008.1 | - | 9 | 1037 | ||
AB025009.1 | - | 8 | 871 | |||
AF290004.1 | - | 12 | 1308 | |||
AK098814.1 | - | 8 | 3928 | |||
AK290702.1 | - | 11 | 1172 | |||
AY789444.1 | - | 11 | 1106 | |||
AY789445.1 | - | 9 | 906 | |||
AY825504.1 | - | 10 | 1067 | |||
AY911310.1 | - | 11 | 1165 | |||
AY911311.1 | - | 10 | 979 | |||
BC036339.1 | - | 9 | 985 | 5184463 | ||
BC047336.1 | - | 10 | 994 | 5185486 | ||
BC106910.1 | - | 8 | 953 | 40031872 | ||
BC139901.1 | - | 10 | 999 | |||
BC139920.1 | - | 10 | 999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
