siRNA Details
Gene Symbol: PKIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270393.1 | NP_001257322.1 | 8 | 877 | ||
NM_001270394.1 | NP_001257323.1 | 8 | 818 | |||
NM_001270395.1 | NP_001257324.1 | 5 | 479 | |||
NM_032471.5 | NP_115860.1 | 4 | 693 | |||
NM_181794.2 | NP_861459.1 | 7 | 757 | |||
NM_181795.2 | NP_861460.1 | 5 | 760 | |||
XM_011535930.2 | XP_011534232.1 | 6 | 870 | |||
XM_011535931.2 | XP_011534233.1 | 7 | 641 | |||
XM_011535932.2 | XP_011534234.1 | 6 | 615 | |||
XM_011535933.2 | XP_011534235.1 | 6 | 576 | |||
XM_011535934.2 | XP_011534236.1 | 4 | 447 | |||
XM_011535935.2 | XP_011534237.1 | 3 | 322 | |||
XM_011535936.1 | XP_011534238.1 | 3 | 323 | |||
XM_011535937.2 | XP_011534239.1 | 3 | 489 | |||
GenBank mRNA | AF087873.1 | - | 5 | 553 | ||
AF225513.1 | - | 3 | 407 | |||
AJ420562.1 | - | 7 | 745 | |||
AK074397.1 | - | 5 | 479 | |||
BC036011.1 | - | 4 | 525 | 4768945 | ||
BC093027.1 | - | 5 | 595 | 30341475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
