siRNA Details
Gene Symbol: PKIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270393.1 | NP_001257322.1 | 8 | 827 | ||
NM_001270394.1 | NP_001257323.1 | 8 | 768 | |||
NM_001270395.1 | NP_001257324.1 | 5 | 429 | |||
NM_032471.5 | NP_115860.1 | 4 | 643 | |||
NM_181794.2 | NP_861459.1 | 7 | 707 | |||
NM_181795.2 | NP_861460.1 | 5 | 710 | |||
XM_011535930.2 | XP_011534232.1 | 6 | 820 | |||
XM_011535931.2 | XP_011534233.1 | 7 | 591 | |||
XM_011535932.2 | XP_011534234.1 | 6 | 565 | |||
XM_011535933.2 | XP_011534235.1 | 6 | 526 | |||
XM_011535934.2 | XP_011534236.1 | 4 | 397 | |||
XM_011535935.2 | XP_011534237.1 | 3 | 272 | |||
XM_011535936.1 | XP_011534238.1 | 3 | 273 | |||
XM_011535937.2 | XP_011534239.1 | 3 | 439 | |||
GenBank mRNA | AF087873.1 | - | 5 | 503 | ||
AF225513.1 | - | 3 | 357 | |||
AJ420562.1 | - | 7 | 695 | |||
AK074397.1 | - | 5 | 429 | |||
BC036011.1 | - | 4 | 475 | 4768945 | ||
BC093027.1 | - | 5 | 545 | 30341475 | ||
BM802552.1 | - | 8 | 788 | |||
BP262756.1 | - | 5 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
