siRNA Details
Gene Symbol: SEMA3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005914.2 | NP_001005914.1 | 18 | 2882 | ||
NM_001290060.1 | NP_001276989.1 | 17 | 2839 | |||
NM_001290061.1 | NP_001276990.1 | 17 | 2688 | |||
NM_001290062.1 | NP_001276991.1 | 11 | 2105 | |||
NM_001290063.1 | NP_001276992.1 | 11 | 1979 | |||
NM_004636.3 | NP_004627.1 | 18 | 2885 | |||
NR_110697.1 | 8 | 1606 | ||||
GenBank mRNA | AB209322.1 | - | 16 | 3065 | ||
AF217991.1 | - | 17 | 2648 | |||
AI309508.1 | - | 1 | 127 | |||
AK092182.1 | - | 8 | 1568 | |||
AK098329.1 | - | 5 | 3171 | |||
BC009113.1 | - | 14 | 2589 | 4153377 | ||
BC013975.2 | - | 17 | 2662 | 4121913 | ||
BC024220.2 | - | 17 | 2662 | 3009807 | ||
BC044602.1 | - | 11 | 2067 | |||
U28369.1 | - | 17 | 2828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
