siRNA Details
Gene Symbol: PREX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024870.3 | NP_079146.2 | 21 | 2587 | ||
NM_025170.5 | NP_079446.3 | 21 | 2587 | |||
XM_011517612.2 | XP_011515914.1 | 21 | 2781 | |||
GenBank mRNA | AJ437636.2 | - | 21 | 2584 | ||
AK023049.1 | - | 16 | 1662 | |||
AK024079.1 | - | 6 | 663 | |||
AK294299.1 | - | 19 | 2018 | |||
AY508996.1 | - | 21 | 2334 | |||
AY508997.1 | - | 21 | 2334 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
