siRNA Details
Gene Symbol: DNAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015512.4 | NP_056327.4 | 30 | 5216 | ||
XM_017006129.1 | XP_016861618.1 | 31 | 8104 | |||
XM_017006130.1 | XP_016861619.1 | 31 | 8103 | |||
XM_017006131.1 | XP_016861620.1 | 31 | 8104 | |||
XM_017006132.1 | XP_016861621.1 | 31 | 8105 | |||
XM_017006133.1 | XP_016861622.1 | 31 | 8105 | |||
GenBank mRNA | AB037831.3 | - | 30 | 5216 | ||
AB290163.1 | - | 30 | 5216 | |||
AJ132083.1 | - | 3 | 329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
