siRNA Details
Gene Symbol: TWIST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000474.3 | NP_000465.1 | 1 | 782 | ||
XM_011515496.1 | XP_011513798.1 | 1 | 529 | |||
GenBank mRNA | BC036704.2 | - | 1 | 782 | 4125830 | |
X99268.1 | - | 1 | 553 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011658, BC083139, BC033434, M63650.
Gene Ontology Categories:
