siRNA Details
Gene Symbol: MTFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003704.2 | NP_001003704.1 | 3 | 994 | ||
NM_016498.4 | NP_057582.2 | 4 | 1227 | |||
GenBank mRNA | AF060924.1 | - | 4 | 1138 | ||
AF151076.1 | - | 4 | 1188 | |||
AK026104.1 | - | 4 | 1095 | |||
BC001608.1 | - | 4 | 1053 | 3542567 | ||
BC009300.2 | - | 4 | 1085 | 4126237 | ||
BC014446.1 | - | 4 | 1053 | 4855192 | ||
BC030989.2 | - | 4 | 1114 | 4077757 | ||
BC038831.2 | - | 4 | 1118 | 3908299 | ||
BC046132.1 | - | 4 | 1017 | 4446034 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
