siRNA Details
Gene Symbol: MTFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003704.2 | NP_001003704.1 | 2 | 430 | ||
NM_016498.4 | NP_057582.2 | 2 | 430 | |||
GenBank mRNA | AF060924.1 | - | 2 | 343 | ||
AF151076.1 | - | 2 | 392 | |||
AK026104.1 | - | 2 | 298 | |||
BC001608.1 | - | 2 | 256 | 3542567 | ||
BC009300.2 | - | 2 | 288 | 4126237 | ||
BC014446.1 | - | 2 | 256 | 4855192 | ||
BC030989.2 | - | 2 | 317 | 4077757 | ||
BC038831.2 | - | 2 | 321 | 3908299 | ||
BC046132.1 | - | 2 | 220 | 4446034 | ||
BQ187590.1 | - | 2 | 212 | |||
CB142469.1 | - | 2 | 321 | |||
CR456345.1 | - | 2 | 225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
