siRNA Details
Gene Symbol: COL18A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030582.3 | NP_085059.2 | 41 | 4549 | ||
NM_130444.2 | NP_569711.2 | 41 | 5254 | |||
NM_130445.3 | NP_569712.2 | 42 | 4082 | |||
GenBank mRNA | AF018081.1 | - | 41 | 4568 | ||
AF018082.1 | - | 42 | 4047 | |||
AF184060.1 | - | 4 | 532 | |||
AF416592.1 | - | 4 | 541 | |||
AK098216.1 | - | 8 | 2172 | |||
AK130835.1 | - | 9 | 1199 | |||
BC033715.1 | - | 30 | 2430 | |||
BC063833.1 | - | 42 | 4023 | 6181818 | ||
L22548.1 | - | 23 | 2032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
