siRNA Details
Gene Symbol: WISP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003880.3 | NP_003871.1 | 4 | 487 | ||
NM_198239.1 | NP_937882.1 | 3 | 567 | |||
NR_125353.1 | 3 | 567 | ||||
NR_125354.1 | 4 | 487 | ||||
XM_011536220.1 | XP_011534522.1 | 4 | 485 | |||
XM_011536223.2 | XP_011534525.1 | 2 | 102 | |||
GenBank mRNA | AB075040.1 | - | 3 | 379 | ||
AF100781.1 | - | 3 | 392 | |||
AF143679.1 | - | 3 | 308 | |||
AY358349.1 | - | 3 | 476 | |||
AY358350.1 | - | 3 | 392 | |||
BC012028.1 | - | 4 | 487 | 4517858 | ||
BC102031.1 | - | 2 | 162 | |||
BC105940.1 | - | 3 | 515 | 40031898 | ||
BC105941.1 | - | 3 | 515 | 40031899 | ||
BG928545.1 | - | 2 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
