siRNA Details
Gene Symbol: ANXA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001155.4 | NP_001146.2 | 16, 17 | 1460 | ||
NM_001193544.1 | NP_001180473.1 | 15, 16 | 1337 | |||
XM_005268432.4 | XP_005268489.1 | 16, 17 | 1460 | |||
GenBank mRNA | AK126836.1 | - | 1445 | |||
AK130077.1 | - | 5, 6 | 301 | |||
AK290672.1 | - | 16, 17 | 1325 | |||
AK298582.1 | - | 15, 16 | 1117 | |||
AK303078.1 | - | 15, 16 | 1323 | |||
BC017046.1 | - | 16, 17 | 1281 | 3912460 | ||
D00510.1 | - | 15, 16 | 1232 | |||
J03578.1 | - | 16, 17 | 1288 | |||
KU177903.1 | - | 15, 16 | 1232 | |||
KU177904.1 | - | 15, 16 | 1232 | |||
Y00097.1 | - | 16, 17 | 1332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
