siRNA Details
Gene Symbol: LRRC27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143757.1 | NP_001137229.1 | 2 | 377 | ||
NM_001143758.1 | NP_001137230.1 | 2 | 224 | |||
NM_001143759.1 | NP_001137231.1 | 2 | 224 | |||
NM_030626.2 | NP_085129.1 | 2 | 224 | |||
XM_006717983.2 | XP_006718046.1 | 2 | 537 | |||
XM_006717984.2 | XP_006718047.1 | 2 | 224 | |||
XM_011540208.1 | XP_011538510.1 | 2 | 307 | |||
XM_011540209.1 | XP_011538511.1 | 2 | 224 | |||
XM_017016690.1 | XP_016872179.1 | 2 | 224 | |||
XM_017016691.1 | XP_016872180.1 | 2 | 224 | |||
XM_017016692.1 | XP_016872181.1 | 2 | 224 | |||
XM_017016693.1 | XP_016872182.1 | 2 | 224 | |||
XM_017016694.1 | XP_016872183.1 | 2 | 170 | |||
GenBank mRNA | AB051461.1 | - | 2 | 134 | ||
AK098652.1 | - | 2 | 180 | |||
AK295953.1 | - | 2 | 141 | |||
BC050097.1 | - | 2 | 179 | 5273395 | ||
DA203895.1 | - | 2 | 377 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
