siRNA Details
Gene Symbol: GRINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000837.1 | NP_000828.1 | 7 | 1920 | ||
NM_001009184.1 | NP_001009184.1 | 7 | 1808 | |||
GenBank mRNA | AB097026.1 | - | 7 | 1732 | ||
AK127640.1 | - | 7 | 1956 | |||
AL157442.1 | - | 1 | 651 | |||
BC041788.1 | - | 7 | 1766 | 4738372 | ||
BC065834.1 | - | 6 | 1674 | |||
BC084553.1 | - | 6 | 1657 | 6206624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
