siRNA Details
Gene Symbol: HSPA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016299.3 | NP_057383.2 | 8 | 849 | ||
GenBank mRNA | AF112210.1 | - | 8 | 668 | ||
AF143723.1 | - | 8 | 658 | |||
AK225265.1 | - | 8 | 693 | |||
AK225651.1 | - | 8 | 693 | |||
AK226065.1 | - | 1 | 3522 | |||
AK292323.1 | - | 8 | 657 | |||
BC119690.1 | - | 8 | 668 | 40116697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
