siRNA Details
Gene Symbol: GPRASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004051.3 | NP_001004051.1 | 5 | 2031 | ||
NM_001184874.2 | NP_001171803.1 | 5 | 2159 | |||
NM_001184875.2 | NP_001171804.1 | 4 | 2071 | |||
NM_001184876.2 | NP_001171805.1 | 5 | 2070 | |||
NM_138437.5 | NP_612446.1 | 4 | 1943 | |||
GenBank mRNA | AK094646.1 | - | 1 | 391 | ||
AK096067.1 | - | 1 | 1279 | |||
AK123832.1 | - | 4 | 1943 | |||
AL832187.1 | - | 1 | 671 | |||
BC013576.1 | - | 1 | 1275 | 3893954 | ||
BC036772.1 | - | 4 | 1707 | 5723418 | ||
BC051707.1 | - | 4 | 2421 |
Gene Symbol: ARMCX5-GPRASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199818.1 | NP_001186747.1 | 7 | 2163 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
