siRNA Details
Gene Symbol: ATP13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020410.2 | NP_065143.2 | 24 | 3335 | ||
GenBank mRNA | AB058728.1 | - | 24 | 3305 | ||
AF288687.1 | - | 24 | 2953 | |||
AK026044.1 | - | 8 | 1485 | |||
AK056420.1 | - | 24 | 3311 | |||
AK074159.1 | - | 15 | 4120 | |||
AK074798.1 | - | 12 | 1840 | |||
AK074920.1 | - | 13 | 1915 | |||
AK095287.1 | - | 7 | 2508 | |||
AK123780.1 | - | 7 | 1679 | |||
AK125861.1 | - | 8 | 3632 | |||
AK130125.1 | - | 15 | 1975 | |||
AK172778.1 | - | 23 | 2766 | |||
AL834180.1 | - | 1 | 1784 | |||
BC009302.2 | - | 6 | 779 | |||
BC069211.1 | - | 5 | 669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
