siRNA Details
Gene Symbol: ATP13A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020410.2 | NP_065143.2 | 23 | 3212 | ||
GenBank mRNA | AB058728.1 | - | 23 | 3182 | ||
AF288687.1 | - | 23 | 2830 | |||
AK026044.1 | - | 7 | 1362 | |||
AK056420.1 | - | 23 | 3188 | |||
AK074159.1 | - | 14 | 3997 | |||
AK074798.1 | - | 11 | 1717 | |||
AK074920.1 | - | 12 | 1792 | |||
AK095287.1 | - | 6 | 2385 | |||
AK123780.1 | - | 6 | 1556 | |||
AK125861.1 | - | 7 | 3509 | |||
AK130125.1 | - | 14 | 1852 | |||
AK172778.1 | - | 22 | 2643 | |||
AL834180.1 | - | 1 | 1442 | |||
BC009302.2 | - | 5 | 656 | |||
BC069211.1 | - | 4 | 546 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
