siRNA Details
Gene Symbol: ARID5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319085.1 | NP_001306014.1 | 7 | 2216 | ||
NM_001319087.1 | NP_001306016.1 | 7 | 2213 | |||
NM_001319092.1 | NP_001306021.1 | 7 | 2385 | |||
NM_001319093.1 | NP_001306022.1 | 6 | 1912 | |||
NM_001319094.1 | NP_001306023.1 | 7 | 2023 | |||
NM_001319096.1 | NP_001306025.1 | 6 | 1915 | |||
NM_212481.2 | NP_997646.1 | 7 | 2054 | |||
XM_011510499.2 | XP_011508801.1 | 7 | 3922 | |||
XM_017003183.1 | XP_016858672.1 | 8 | 2559 | |||
XM_017003184.1 | XP_016858673.1 | 6 | 2040 | |||
XM_017003185.1 | XP_016858674.1 | 6 | 2271 | |||
GenBank mRNA | AK300173.1 | - | 8 | 1837 | ||
AK307768.1 | - | 7 | 2023 | |||
BC047390.1 | - | 7 | 2388 | |||
BC067301.1 | - | 6 | 1862 | |||
BX648918.1 | - | 6 | 1883 | |||
M62324.1 | - | 7 | 2006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
