siRNA Details
Gene Symbol: GJA9-MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037633.1 | 3, 4 | 2870 | |||
NR_037634.1 | 4, 5 | 2002 | ||||
NR_037635.1 | 2, 3 | 271 | ||||
GenBank mRNA | AK123051.1 | - | 4, 5 | 1996 | ||
BC039090.1 | - | 2, 3 | 891 | |||
BC051675.1 | - | 3, 4 | 2842 | |||
FY211295.1 | - | 2, 3 | 247 |
Gene Symbol: MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012333.4 | NP_036465.2 | 2, 3 | 164 | ||
GenBank mRNA | AB007191.2 | - | 2, 3 | 129 | ||
AB451287.1 | - | 2, 3 | 91 | |||
AB451419.1 | - | 2, 3 | 91 | |||
AK311886.1 | - | 2, 3 | 154 | |||
BC008686.1 | - | 2, 3 | 129 | 3870100 | ||
BT007109.1 | - | 2, 3 | 91 | |||
BU601195.1 | - | 2, 3 | 166 | |||
CR456936.1 | - | 2, 3 | 91 | |||
D50692.1 | - | 2, 3 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
